Order Entry
Puerto Rico
ContactUsLinkComponent
PRIME-XV IS21 Supplement (50X)
PRIME-XV IS21 Supplement (50X)
Catalog # 77975-332
New Product
CAS Number:  
undefined
PRIME-XV IS21 Supplement (50X)
Catalog # 77975-332
Supplier Number:  91142-10ML
New Product
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pack type:
    Bottle
  • Size:
    10 ml
  • Shelf life:
    24
  • Application:
    For long-term neuronal cultures

Specifications

About this item

Neural supplement for long-term neuronal cultures.

  • Neural supplement for use with PRIME-XV Neural Basal

    Medium and conventional media such as Ham’s DMEM/F12 with L-glutamine

    Does not contain antibiotics

PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long-term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be diluted to 1X in basal medium before use. This supplement does not contain antibiotics. Designed and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.

Certifications: FDA, Federal, and State registered - cGMP-compliant manufacture; EN ISO 13485:2016 certified; MDSAP certified; Extensive QC testing including functionality, sterility (USP <71>), and endotoxin (USP <85>); Drug Master Files (DMFs) filed with the FDA – available upon request.