Ham's F12K (Kaighn's Nutrient Mixture F12) was originally developed for use in cloning Chinese hamster ovary and lung cells. This medium is useful with serum supplements for a variety of cell culture applications, including the growth of myeloma and hybridoma cells.
- Animal component-free
- Chemically defined
- GMP manufacture
With:
2500 mg/L Sodium bicarbonate
Without:
Ascorbic acid
Linoleic acid
L-Glutamine - add 10 ml/L (200 mM)
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Calcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:Yes
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Low
- Media Base:Ham's F12K (Kaighn's) Nutrient Mixture