Order Entry
Puerto Rico
ContactUsLinkComponent
PRIME-XV MSC XSFM Dual Component Kit without Phenol Red
PRIME-XV MSC XSFM Dual Component Kit without Phenol Red
Catalog # 77975-347
New Product
CAS Number:  
undefined
PRIME-XV MSC XSFM Dual Component Kit without Phenol Red
Catalog # 77975-347
Supplier Number:  91214DC
New Product
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pack type:
    Bag
  • Size:
    5 L basal media and 10 ml supplement
  • Calcium:
    No
  • Magnesium:
    No
  • HEPES:
    No
  • Sodium bicarbonate:
    No
  • Sodium pyruvate:
    No
  • Stable glutamine:
    No
  • L-Glutamine:
    No
  • Glucose level:
    Without
  • Media Base:
    Proprietary mixture for human primary cells
  • Shelf life:
    12

Specifications

About this item

The PRIME-XV MSC XSFM Dual Component Kits are xeno-free and serum-free media that enable a single-use, closed system format for MSC expansion in Quantum or other compatible hollow fiber bioreactors. The medium is formulated to deliver expansion and viability of MSCs over several passages in culture, while maintaining multipotency and functional characteristics.

  • Enables up to an eight-fold increase in MSC expansion (donor-dependent)
  • Yields 200×106 cells in seven days of expansion with 90% and above viability
  • Supports expansion of MSCs; data derived from human adipose tissue
  • Maintains lot-to-lot consistency with xeno- and serum-free media
  • Simplifies and expedites process of MSC expansion for large scale solutions
  • Minimizes risk of contamination and cell stress, and increases efficiency
  • Does not contain antibiotics
  • Available with or without phenol red

PRIME-XV MSC XSFM Dual component kit with phenol red, 91149DC is a dual-component serum-free and xeno-free 5 L basal medium bag and a frozen supplement bag. When combined, the formulation is optimized for the maintenance and expansion of purified or enriched human mesenchymal stromal/stem cells (MSCs) in the Terumo Quantum or Quantum Flex bioreactor, or other compatible bioreactors, under serum-free and xeno-free culture conditions. This product does not contain antibiotics. This medium is provided as a basal medium and supplement and should be mixed prior to use. It may also be used with additional cytokine/growth factors for desired application.