HBSS 10X-Hanks' Balanced Salt Solution
Catalog # 77975-368
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
New Product
CAS Number:
Specifications
- Size:500 ml
- Calcium:No
- Sodium bicarbonate:No
- Phenol red:Yes
- Format:Liquid
Specifications
About this item
HBSS 10X Hanks' Balanced salt solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions. HBSS is suitable for use in sealed culture flasks incubated in air.
- Phenol red
Packaging:
Store at 15 to 30 °C and protect from light
Shelf life: 24 months from date of manufacture