BalanCD Transfectory CHO system of media and supplements is a scalable platform that produces milligram to gram-scale yields of recombinant proteins using a simplified, affordable protocol. The system (including BalanCD Transfectory CHO, transfectory supplement and anti-clumping supplement) supports small- and large-scale transient transfection in CHO cells, enabling scientists to use the same medium before and after transfection. The chemically-defined, animal component-free medium produces protein levels equivalent to, or better than, other commercially available media.
Scalable protocol eliminates need to exchange medium before or after transfection
Quickly achieves high protein yields
Saves time and reduces development costs vs conventional stable cell line development
Compatible with most commercially available transfection reagents (e. g. polyethyleneimine [PEI])
Rapid, scalable, high yield protein production
BalanCD Transfectory CHO System. Rapid, scalable, high yield protein production. The BalanCD Transfectory CHO system of medium and supplements is a scalable platform that produces milligram to gram-scale yields of recombinant proteins using a simplified, affordable protocol. The system supports small- and large-scale transient transfection in CHO cells enabling scientists to work with CHO cells from early phase discovery through to production. BalanCD Transfectory CHO is a chemically defined, animal derived component free medium optimized for small- and large-scale transient transfection and protein production in CHO cells grown in suspension. The medium contains no hydrolysates or any other undefined components. This medium is supplied ready to use for suspension culture applications.
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Phenol red:No
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Without
- Media Base:Proprietary Mixture