Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Media Format:Liquid
- Size:500 ml
- Calcium:No
- Magnesium:No
- HEPES:No
- Sodium bicarbonate:No
- Sodium pyruvate:No
- Phenol red:No
- Stable glutamine:No
- L-Glutamine:No
- Glucose level:Without
- Media Base:AMEM/Alpha - Minimum Essential Medium Alpha Modification
Specifications
About this item
Alpha MEM Earle's salts without nucleosides is modified Eagle's minimum essential medium designed for growth of a mouse-hamster-hybridoma cell line (8-A).
- Animal-component free
- Chemically defined
- GMP manufacture
Alpha-MEM is a modified Eagle’s MEM that includes Earle’s Balanced Salts. When supplemented with serum or serum substitutes, Alpha-MEM is suitable, and commonly used, for culturing bone marrow- and amniotic fluid-derived cells, including progenitor cells.