Order Entry
Puerto Rico
ContactUsLinkComponent
Hardness QC Certified Reference Material
Hardness QC Certified Reference Material
Catalog # SXQCI-137
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
Hardness QC Certified Reference Material
Catalog # SXQCI-137
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  QCI-137
New Product
CAS Number:  

Specifications

  • Concentration:
    Multiple
  • Description:
    Hardness QC certified reference material 250 ml
  • Shipping Conditions:
    Non-hazardous
  • Physical Form:
    Liquid
  • Contents:
    Multiple

Specifications

About this item

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF and other analytical instrumentation

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

For US EPA Methods 130.1, 130.2, 160.2, 200.7, 200.8, 215.1, 215.2, 242.1, 6010, 6010B, 6020, 7140, 7450; SM 18/19th ED 2340 B; SM 18/19th ED 2340 C; SM 18/19th ED 3500 Ca-D; SM 18/19th ED 3500-Mg E; SM 18th ED 2540 D; SM 18th ED 3500-Ca D; SM 18th ED 3500-Mg D; and SM 3120.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016.