Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acidity QC Concentrate, Certified Reference Material

Acidity QC Concentrate, Certified Reference Material

Acidity QC concentrate, certified reference material 125 ml, 650 - 1800 µg/ml, Acidity as CaCO₃
Catalog # SXQCI-099
Supplier:   SPEX CERTIPREP LLC
Supplier #:  QCI-099
Acidity QC Concentrate, Certified Reference Material
Catalog # SXQCI-099
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Acidity QC Concentrate, Certified Reference Material
Acidity QC Concentrate, Certified Reference Material

Acidity QC concentrate, certified reference material 125 ml, 650 - 1800 µg/ml, Acidity as CaCO₃

CAS Number:  
Price
$62.81
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation For US EPA Method 305.1 and SM 18th ED 2310 B (4A)

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016.

Specifications

Specifications

  • Concentration:
    650 - 1800 µg/ml
  • Description:
    Acidity QC concentrate, certified reference material 125 ml
  • Contents:
    Acidity as CaCO₃
  • Cat. No.:
    SXQCI-099
  • Physical Form:
    Liquid
  • Shipping Conditions:
    Non-Hazardous
  • Storage Temperature:
    Ambient

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.