Order Entry
Puerto Rico
ContactUsLinkComponent
Trihalomethanes QC Concentrate
Trihalomethanes QC Concentrate
Catalog # NSQCO-002
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
undefined
Trihalomethanes QC Concentrate
Catalog # NSQCO-002
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  QCO-002
CAS Number:  

Specifications

  • Concentration:
    5 to 50 µg/L
  • Description:
    Trihalomethanes QC concentrate, 1.5 ml
  • Shipping Conditions:
    Non-hazardous
  • Storage Temperature:
    Ambient
  • Physical Form:
    Liquid
  • Contents:
    Bromodichloromethane, Bromoform, Chlorodibromomethane, Chloroform
  • Contents:
    Bromodichloromethane, Bromoform, Chlorodibromomethane, Chloroform

Specifications

About this item

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation

    For US EPA Methods 502.1, 502.2, 503.1, 524.1, 524.2, and 551

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016