Order Entry
Puerto Rico
ContactUsLinkComponent
Residual Free Chlorine QC Concentrate, Certified Reference Material
Residual Free Chlorine QC Concentrate, Certified Reference Material
Catalog # NSQCI-012
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
undefined
Residual Free Chlorine QC Concentrate, Certified Reference Material
Catalog # NSQCI-012
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  QCI-012
CAS Number:  

Specifications

  • Concentration:
    0.5 to 3 µg/ml
  • Description:
    Residual free chlorine QC concentrate, certified reference material, 2.2 ml
  • Shipping Conditions:
    Non-hazardous
  • Storage Temperature:
    Ambient
  • Physical Form:
    Liquid
  • Contents:
    Free Residual Chlorine, Total Residual Chlorine
  • Contents:
    Free Residual Chlorine, Total Residual Chlorine

Specifications

About this item

Meet or exceed performance criteria with QC samples.

  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation

    For US EPA Methods 330.1, 330.2, 330.3, 330.4, 330.5; SM 18/19th ED 4500 Cl- B; SM 18/19th ED 4500 Cl- D; SM 18/19th ED 4500-Cl E; SM 18/19th ED 4500-Cl F; SM 18/19th ED 4500-Cl G; SM 18/19th ED 4500-Cl H; SM 18/19th ED 4500-Cl I; SM 18th ED 4500 Cl B; SM 18th ED 4500 Cl C; SM 18th ED 4500 Cl- E; SM 18th ED 4500-Cl D; SM 18th ED 4500-Cl F; and SM 18th ED 4500-Cl G

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016