Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Chondroitin Sulfate Mouse Monoclonal Antibody [clone: CS-56]
Anti-Chondroitin Sulfate Mouse Monoclonal Antibody [clone: CS-56]
Catalog # ABCA_AB11570-50UL
Supplier:  Abcam
New Product
Anti-Chondroitin Sulfate Mouse Monoclonal Antibody [clone: CS-56]
Catalog # ABCA_AB11570-50UL
Supplier:  Abcam
Supplier Number:  AB11570-50UL
New Product

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    0
  • Antigen Symbol:
    Chondroitin Sulfate
  • Clonality:
    Monoclonal
  • Clone:
    CS-56
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgM
  • Size:
    50 µl
  • Form:
    Liquid
  • Storage Buffer:
    Potassium phosphate, Sodium chloride
  • Storage Temperature:
    Upon delivery aliquot and Store at –20 or –80 °C. Avoid repeated freeze/thaw cycles.
  • Shipping Temperature:
    Shipped at +4 °C
  • Tested Applications:
    EM, ICC/IF, IHC-P
  • Purification:
    Proprietary technique
  • Packaging:
    Vial

Specifications

About this item

Mouse monoclonal [CS-56] to Chondroitin Sulfate.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Chondroitin Sulfate
Clonality: Monoclonal
Clone: CS-56
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: