Anti-Cell adhesion molecule 2 Rabbit Polyclonal Antibody
ABCA_AB64873-100UG
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:IGSF4D
- Antigen Symbol:Cell adhesion molecule 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:NECL3|CADM2|Immunoglobulin superfamily member 4D|Nectin-like protein 3|Synaptic cell adhesion molecule 2|NECL-3|SynCAM 2
- Storage Buffer:PBS, Glycerol, Sodium chloride
- Molecular Weight:47554 Da
- Storage Temperature:Shipped at +4 °C. Store at –20 °C. Stable for 12 months at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Synthetic Peptide within Human CADM2. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to Cell adhesion molecule 2.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cell adhesion molecule 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human