Anti-POT1 Rabbit Monoclonal Antibody [clone: EPR25161-142]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR25161-142] to POT1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: POT1
Clonality: Monoclonal
Clone: EPR25161-142
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB309475-100U
- ABCA_AB309475-20UL
- ABCA_AB309475-1ML
- Antigen Symbol:
- POT1
- POT1
- POT1
- Antigen Name:
- Protection of telomeres protein 1
- Protection of telomeres protein 1
- Protection of telomeres protein 1
- Antigen Synonyms:
- hPot1|POT1-like telomere end-binding protein
- hPot1|POT1-like telomere end-binding protein
- hPot1|POT1-like telomere end-binding protein
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR25161-142
- EPR25161-142
- EPR25161-142
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 71442 Da
- 71442 Da
- 71442 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipping Temperature:
- +4 °C
- +4 °C
- +4 °C
- Tested Applications:
- IP
- IP
- IP