Anti-Cathepsin L/V/K/H Rabbit Monoclonal Antibody [clone: EPR8011] (Alexa Fluor® 568)
Catalog # ABCA_AB312354-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:CTSL1
- Antigen Symbol:Cathepsin L/V/K/H
- Clonality:Monoclonal
- Clone:EPR8011
- Conjugation:Alexa Fluor® 568
- Host:Rabbit
- Isotype:IgG
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:CTSL|Procathepsin L|Cathepsin L1|Major excreted protein|MEP
- Molecular Weight:37564 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:Antibody labelling, target binding affinity
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Alexa Fluor® 568 Rabbit monoclonal [EPR8011] to Cathepsin L/V/K/H.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cathepsin L/V/K/H
Clonality: Monoclonal
Clone: EPR8011
Conjugation: Alexa Fluor® 568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: