Anti-Alpha-synuclein Mouse Monoclonal Antibody [clone: LB 509]
Catalog # ABCA_AB264092-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:NACP
- Antigen Symbol:Alpha-synuclein
- Clonality:Monoclonal
- Clone:LB 509
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Epitope:ab27766 reacts with an epitope located in the region encoded by amino acids 115-122 of alpha-synuclein.
- Form:Liquid
- Antigen Synonyms:PARK1|SNCA|Non-A beta component of AD amyloid|Non-A4 component of amyloid precursor
- Molecular Weight:14460 Da
- Storage Temperature:Store at +4 °C. Do not freeze.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:Flow Cyt (Intra)
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [LB 509] to Alpha-synuclein - BSA and Azide free.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Alpha-synuclein
Clonality: Monoclonal
Clone: LB 509
Conjugation:
Epitope: ab27766 reacts with an epitope located in the region encoded by amino acids 115-122 of alpha-synuclein.
Host: Mouse
Isotype: IgG1
Reactivity: Human; Rat