Anti-Proteasome subunit beta type 2/PSMB2 Rabbit Monoclonal Antibody [clone: EPR8827]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR8827] to Proteasome subunit beta type 2/PSMB2 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Proteasome subunit beta type 2/PSMB2
Clonality: Monoclonal
Clone: EPR8827
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB248824-100U
- ABCA_AB248824-1MG
- Antigen Symbol:
- Proteasome subunit beta type 2/PSMB2
- Proteasome subunit beta type 2/PSMB2
- Antigen Name:
- Proteasome subunit beta type-2
- Proteasome subunit beta type-2
- Antigen Synonyms:
- Macropain subunit C7-I|Multicatalytic endopeptidase complex subunit C7-I|Proteasome component C7-I|Proteasome subunit beta-4|beta-4|PSMB2
- Macropain subunit C7-I|Multicatalytic endopeptidase complex subunit C7-I|Proteasome component C7-I|Proteasome subunit beta-4|beta-4|PSMB2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR8827
- EPR8827
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 22836 Da
- 22836 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P