Anti-CXCL10 Rabbit Recombinant Multiclonal Antibody [clone: RM2054]
: Abcam
New Product
Rabbit recombinant multiclonal [RM2054] to CXCL10.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CXCL10
Clonality: Recombinant Multiclonal
Clone: RM2054
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB318282-100U
- ABCA_AB318282-20UL
- ABCA_AB318282-1ML
- Antigen Symbol:
- CXCL10
- CXCL10
- CXCL10
- Antigen Name:
- INP10
- INP10
- INP10
- Antigen Synonyms:
- SCYB10|C-X-C motif chemokine 10|10 kDa interferon gamma-induced protein|Small-inducible cytokine B10|Gamma-IP10|IP-10
- SCYB10|C-X-C motif chemokine 10|10 kDa interferon gamma-induced protein|Small-inducible cytokine B10|Gamma-IP10|IP-10
- SCYB10|C-X-C motif chemokine 10|10 kDa interferon gamma-induced protein|Small-inducible cytokine B10|Gamma-IP10|IP-10
- Antibody Type:
- Primary
- Primary
- Primary
- Clone:
- RM2054
- RM2054
- RM2054
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 10881 Da
- 10881 Da
- 10881 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P