Anti-SUN2 Rabbit Polyclonal Antibody
Catalog # ABCA_AB87036-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Unc84b
- Antigen Symbol:SUN2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:SUN domain-containing protein 2|Protein unc-84 homolog B|Sad1/unc-84 protein-like 2
- Storage Buffer:Whole serum
- Molecular Weight:81605 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid repeated freeze/thaw cycles.
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Mouse Sun2 aa 1 - 50. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC
- Purification:Whole antiserum
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to SUN2.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SUN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse