Anti-ZNF638 Rabbit Polyclonal Antibody
ABCA_AB183771-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:NP220
- Antigen Symbol:ZNF638
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:ZFML|Zinc finger protein 638|Cutaneous T-cell lymphoma-associated antigen se33-1|Nuclear protein 220|Zinc finger matrin-like protein|CTCL-associated antigen se33-1
- Molecular Weight:220625 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Recombinant Fragment Protein within Human ZNF638 aa 1 - 350. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to ZNF638.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ZNF638
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human