Anti-BubR1 (phospho T680) Rabbit Monoclonal Antibody [clone: EPR19958]
: Abcam
New Product
Rabbit monoclonal [EPR19958] to BubR1 (phospho T680).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: BubR1
Clonality: Monoclonal
Clone: EPR19958
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB200061-1ML
- ABCA_AB200061-100U
- Antigen Symbol:
- BubR1
- BubR1
- Antigen Name:
- MAD3L
- MAD3L
- Antigen Synonyms:
- SSK1|BUB1B|Mitotic checkpoint serine/threonine-protein kinase BUB1 beta|MAD3/BUB1-related protein kinase|Mitotic checkpoint kinase Protein SSK1|hBUBR1
- SSK1|BUB1B|Mitotic checkpoint serine/threonine-protein kinase BUB1 beta|MAD3/BUB1-related protein kinase|Mitotic checkpoint kinase Protein SSK1|hBUBR1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR19958
- EPR19958
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Modification:
- phospho T680
- phospho T680
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 119545 Da
- 119545 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipping Temperature:
- +4 °C
- +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF
- Flow Cyt (Intra), ICC/IF
Frequently Bought Together