Anti-MPO Rat Monoclonal Antibody [clone: EPR20257]
Supplier: Abcam
New Product
Certificates
About this item
Rat monoclonal [EPR20257] to Myeloperoxidase - Chimeric.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: EPR20257
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB300650-100U
- ABCA_AB300650-1ML
- ABCA_AB300650-20UL
- Antigen Symbol:
- MPO
- MPO
- MPO
- Antigen Name:
- Myeloperoxidase
- Myeloperoxidase
- Myeloperoxidase
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR20257
- EPR20257
- EPR20257
- Host:
- Rat
- Rat
- Rat
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 83869 Da
- 83869 Da
- 83869 Da
- Purification:
- Ion exchange chromatography
- Ion exchange chromatography
- Ion exchange chromatography
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P