Anti-Src Mouse Monoclonal Antibody [clone: Clone 327]
Catalog # ABCA_AB16885-50UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:SRC1
- Antigen Symbol:Src
- Clonality:Monoclonal
- Clone:Clone 327
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western Blot:Yes
- Size:50 µg
- Epitope:Epitope is within the SH3 domain of either v-Src or c-Src. We have no evidence for ab16885 cross-reacting with other src family kinases (e.g.fyn)
- Form:Liquid
- Antigen Synonyms:Proto-oncogene tyrosine-protein kinase Src|Proto-oncogene c-Src|pp60c-src|p60-Src
- Storage Buffer:Sodium phosphate, gelatin
- Molecular Weight:59835 Da
- Storage Temperature:Store at 4 °C. Do not freeze.
- Concentration:0.1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Native Full Length Protein corresponding to Human SRC
- Purification:Tissue culture supernatant
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [Clone 327] to Src.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Src
Clonality: Monoclonal
Clone: Clone 327
Conjugation:
Epitope: Epitope is within the SH3 domain of either v-Src or c-Src. We have no evidence for ab16885 cross-reacting with other src family kinases (e.g.fyn)
Host: Mouse
Isotype: IgG1
Reactivity: Human