Anti-Chemerin Rabbit Polyclonal Antibody
ABCA_AB103153-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:TIG2
- Antigen Symbol:Chemerin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RARRES2|Retinoic acid receptor responder protein 2|RAR-responsive protein Tazarotene-induced gene 2 protein
- Storage Buffer:PBS
- Molecular Weight:18618 Da
- Storage Temperature:Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Native Full Length Protein corresponding to Human RARRES2.
- Purification:Protein A Affinity purification
- Packaging:Vial
Rabbit polyclonal to Chemerin.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Chemerin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse