Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: 11-B3]
Catalog # ABCA_AB18929-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Genome polyprotein
- Antigen Symbol:Hepatitis C Virus Core Antigen
- Clonality:Monoclonal
- Clone:11-B3
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Epitope:Amino acids 70-90
- Form:Liquid
- Storage Buffer:PBS
- Molecular Weight:327051 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 or −80 °C. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Tested Applications:IHC-Fr
- Purification:Protein G affinity purification
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [11-B3] to Hepatitis C Virus Core Antigen.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Hepatitis C Virus Core Antigen
Clonality: Monoclonal
Clone: 11-B3
Conjugation:
Epitope: Amino acids 70-90
Host: Mouse
Isotype: IgG1
Reactivity: HCV