Anti-non-muscle Myosin IIA Rabbit Monoclonal Antibody [clone: EPR8965]
: Abcam
New Product
Rabbit monoclonal [EPR8965] to non-muscle Myosin IIA.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: non-muscle Myosin IIA
Clonality: Monoclonal
Clone: EPR8965
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB138498-1ML
- ABCA_AB138498-100U
- Antigen Synonyms:
- Myosin heavy chain 9|Non-muscle myosin heavy chain A|Non-muscle myosin heavy chain IIa|NMMHC-A|NMMHC II-a|NMMHC-IIA|MYH9
- Myosin heavy chain 9|Non-muscle myosin heavy chain A|Non-muscle myosin heavy chain IIa|NMMHC-A|NMMHC II-a|NMMHC-IIA|MYH9
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 226532 Da
- 226532 Da
- Purification:
- Tissue culture supernatant
- Tissue culture supernatant
- Storage Temperature:
- +4 °C
- +4 °C
- Shipping Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze / thaw cycle.
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P