Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TNF Receptor I Rabbit Monoclonal Antibody [clone: EPR23742-65]
Anti-TNF Receptor I Rabbit Monoclonal Antibody [clone: EPR23742-65]
Supplier:  Abcam
New Product
Certificates
Anti-TNF Receptor I Rabbit Monoclonal Antibody [clone: EPR23742-65]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB259817-100U
  • ABCA_AB259817-1ML
  • ABCA_AB259817-20UL
  • Antigen Symbol:
  • TNF Receptor I
  • TNF Receptor I
  • TNF Receptor I
  • Antigen Name:
  • CD120a
  • CD120a
  • CD120a
  • Antigen Synonyms:
  • TNFAR|TNFR1|TNFRSF1A|Tumor necrosis factor receptor superfamily member 1A|Tumor necrosis factor receptor 1|Tumor necrosis factor receptor type I|p55|p60|TNF-R1|TNF-RI|TNFR-I
  • TNFAR|TNFR1|TNFRSF1A|Tumor necrosis factor receptor superfamily member 1A|Tumor necrosis factor receptor 1|Tumor necrosis factor receptor type I|p55|p60|TNF-R1|TNF-RI|TNFR-I
  • TNFAR|TNFR1|TNFRSF1A|Tumor necrosis factor receptor superfamily member 1A|Tumor necrosis factor receptor 1|Tumor necrosis factor receptor type I|p55|p60|TNF-R1|TNF-RI|TNFR-I
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR23742-65
  • EPR23742-65
  • EPR23742-65
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 50495 Da
  • 50495 Da
  • 50495 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • 4 °C

Specifications