Anti-ATG12 Rabbit Recombinant Multiclonal Antibody [clone: RP23040104]
Catalog # ABCA_AB313434-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:APG12
- Antigen Symbol:ATG12
- Clonality:Recombinant Multiclonal
- Clone:RP23040104
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:APG12L|Ubiquitin-like protein ATG12|Autophagy-related protein 12|APG12-like
- Molecular Weight:15113 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human ATG12
- Tested Applications:ICC/IF
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit recombinant multiclonal [RP23040104] to ATG12.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ATG12
Clonality: Recombinant Multiclonal
Clone: RP23040104
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse