Anti-Prostaglandin E Receptor EP1/PTGER1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB183073-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Prostaglandin E2 receptor EP1 subtype
- Antigen Symbol:Prostaglandin E Receptor EP1/PTGER1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:PGE receptor EP1 subtype|PGE2 receptor EP1 subtype|Prostanoid EP1 receptor|PTGER1
- Molecular Weight:41801 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:0.9 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PTGER1 aa 350 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Prostaglandin E Receptor EP1/PTGER1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Prostaglandin E Receptor EP1/PTGER1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human