Anti-MTCO2 Mouse Monoclonal Antibody [clone: 12C4F12]
ABCA_AB110258-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:COII
- Antigen Symbol:MTCO2
- Clonality:Monoclonal
- Clone:12C4F12
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:COX2|COXII|MT-CO2|Cytochrome c oxidase subunit 2|Cytochrome c oxidase polypeptide II
- Molecular Weight:25565 Da
- Storage Temperature:Store at 4 °C
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:ICC/IF
- Purification:IgG fraction
- Packaging:Vial
Mouse monoclonal [12C4F12] to MTCO2.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MTCO2
Clonality: Monoclonal
Clone: 12C4F12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human