Anti-Smad2+Smad3 Mouse Monoclonal Antibody [clone: 18-Smad2-3]
: Abcam
New Product
Mouse monoclonal [18/Smad2/3] to Smad2 + Smad3 - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Smad2+Smad3
Clonality: Monoclonal
Clone: 18-Smad2-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat
- Catalog No:
- ABCA_AB305326-1MG
- ABCA_AB305326-100U
- Antigen Symbol:
- Smad2+Smad3
- Smad2+Smad3
- Antigen Name:
- Madh2
- Madh2
- Antigen Synonyms:
- Madr2|Smad2|Mothers against decapentaplegic homolog 2|MAD homolog 2|Mothers against DPP homolog 2|Mad-related protein 2|SMAD family member 2|mMad2|SMAD 2
- Madr2|Smad2|Mothers against decapentaplegic homolog 2|MAD homolog 2|Mothers against DPP homolog 2|Mad-related protein 2|SMAD family member 2|mMad2|SMAD 2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 18-Smad2-3
- 18-Smad2-3
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 52266 Da
- 52266 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P
- IHC-P