Anti-SOX11 Mouse Monoclonal Antibody [clone: CL0143]
Catalog # ABCA_AB242203-100U
Supplier: Abcam
New Product
Specifications
- Size:100 µl
- Packaging:Vial
Specifications
About this item
Mouse monoclonal [CL0143] to SOX11.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SOX11
Clonality: Monoclonal
Clone: CL0143
Conjugation:
Epitope: Binds to an epitope located within the peptide sequence IPFIREAERL as determined by overlapping synthetic peptides.
Host: Mouse
Isotype: IgG2a
Reactivity: Human