Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-C3orf43 Mouse Monoclonal Antibody [clone: 1H6]
Anti-C3orf43 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # ABCA_AB230681-50UL
Supplier:  Abcam
New Product
Anti-C3orf43 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # ABCA_AB230681-50UL
Supplier:  Abcam
Supplier Number:  AB230681-50UL
New Product

About this item

Mouse monoclonal [1H6] to C3orf43.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: C3orf43
Clonality: Monoclonal
Clone: 1H6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human