Anti-TLE 1 Rabbit Monoclonal Antibody [clone: EPR9386(2)]
: Abcam
New Product
Rabbit monoclonal [EPR9386(2)] to TLE 1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TLE 1
Clonality: Monoclonal
Clone: EPR9386(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB183742-1ML
- ABCA_AB183742-100U
- Antigen Synonyms:
- E(Sp1) homolog|Enhancer of split groucho-like protein 1|ESG1|TLE1
- E(Sp1) homolog|Enhancer of split groucho-like protein 1|ESG1|TLE1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 83201 Da
- 83201 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Temperature:
- +4 °C
- +4 °C
- Shipping Temperature:
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Shipped at +4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Tested Applications:
- ICC/IF, IHC-P
- ICC/IF, IHC-P