Anti-Laminin alpha 5/LAMA5 Mouse Monoclonal Antibody [clone: CL3118]
ABCA_AB210957-100U
: Abcam
New Product
- Size:100 µl
- Packaging:Vial
Mouse monoclonal [CL3118] to Laminin alpha 5/LAMA5.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Laminin alpha 5/LAMA5
Clonality: Monoclonal
Clone: CL3118
Conjugation:
Epitope: ab210957 binds to an epitope located within the peptide sequence RQGLVDRAQQ (aa 2555 - 2564).
Host: Mouse
Isotype: IgG1
Reactivity: Human