Anti-PPP2R3A Rabbit Polyclonal Antibody
Catalog # ABCA_AB218165-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:PPP2R3
- Antigen Symbol:PPP2R3A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit alpha|PP2A subunit B isoform PR72/PR130|PP2A subunit B isoform R3 isoform|PP2A subunit B isoforms B''-PR72/PR130|PP2A subunit B isoforms B72/B130|Serine/threonine-protein phosphatase 2A 72/130 kDa regulatory subunit B
- Molecular Weight:130278 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:4 °C
- Immunogen:Synthetic Peptide within Human PPP2R3A aa 800-900 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to PPP2R3A.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: PPP2R3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat