Anti-Phospholipase C gamma 1/PLC-gamma-1 (phospho Y783) Rabbit Monoclonal Antibody [clone: EP1898Y]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EP1898Y] to Phospholipase C gamma 1/PLC-gamma-1 (phospho Y783).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Phospholipase C gamma 1/PLC-gamma-1
Clonality: Monoclonal
Clone: EP1898Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Specifications
- Catalog No:
- ABCA_AB76031-100UL
- ABCA_AB76031-1ML
- Antigen Symbol:
- Phospholipase C gamma 1/PLC-gamma-1
- Phospholipase C gamma 1/PLC-gamma-1
- Antigen Name:
- PLC1
- PLC1
- Antigen Synonyms:
- PLCG1|PLC-148|Phosphoinositide phospholipase C-gamma-1|Phospholipase C-II|Phospholipase C-gamma-1|PLC-II|PLC-gamma-1
- PLCG1|PLC-148|Phosphoinositide phospholipase C-gamma-1|Phospholipase C-II|Phospholipase C-gamma-1|PLC-II|PLC-gamma-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EP1898Y
- EP1898Y
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Modification:
- phospho Y783
- phospho Y783
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 148532 Da
- 148532 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- Storage Temperature:
- Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Dot, ICC/IF
- Dot, ICC/IF