Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y] (Alexa Fluor® 405)
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y] (Alexa Fluor® 405)
Catalog # ABCA_AB210143-100U
Supplier:  Abcam
New Product
undefined
Anti-Sodium Potassium ATPase Rabbit Monoclonal Antibody [clone: EP1845Y] (Alexa Fluor® 405)
Catalog # ABCA_AB210143-100U
Supplier:  Abcam
Supplier Number:  AB210143-100UL
New Product

Specifications

  • Antibody Type:
    Control
  • Antigen Name:
    Sodium/potassium-transporting ATPase subunit alpha-1
  • Antigen Symbol:
    Sodium Potassium ATPase
  • Clonality:
    Monoclonal
  • Clone:
    EP1845Y
  • Conjugation:
    Alexa Fluor® 405
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    Na(+)/K(+) ATPase alpha-1 subunit|Sodium pump subunit alpha-1|ATP1A1
  • Molecular Weight:
    112896 Da
  • Storage Temperature:
    Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C. Avoid freeze/thaw cycle. Store in the dark.
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    Shipped at +4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information.
  • Tested Applications:
    Flow Cyt (Intra)
  • Purification:
    Protein A Affinity purification
  • Packaging:
    Vial

Specifications

About this item

Alexa Fluor® 405 Rabbit monoclonal [EP1845Y] to Sodium Potassium ATPase - Plasma Membrane Loading Control.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EP1845Y
Conjugation: Alexa Fluor® 405
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human