Anti-Histone H2A Rabbit Polyclonal Antibody
Catalog # ABCA_AB18255-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:H2AFM
- Antigen Symbol:Histone H2A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- ChIP:Yes
- Form:Liquid
- Antigen Synonyms:HIST1H2AB|H2AC8|H2AFA|HIST1H2AE|H2AC4|Histone H2A type 1-B/E|Histone H2A.2|Histone H2A/a|Histone H2A/m
- Storage Buffer:PBS, BSA
- Molecular Weight:14135 Da
- Storage Temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 or –80 °C. Avoid freeze/thaw cycle.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:ICC/IF
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to Histone H2A - ChIP Grade.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human; Bovine