Anti-Histone H3.3 Rabbit Monoclonal Antibody [clone: EPR23526-89]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR23526-89] to Histone H3.3 (mutated K36M) - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3.3
Clonality: Monoclonal
Clone: EPR23526-89
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB277487-1MG
- ABCA_AB277487-100U
- Antigen Symbol:
- Histone H3.3
- Histone H3.3
- Antigen Name:
- H3.3A
- H3.3A
- Antigen Synonyms:
- H3F3|H3F3A|PP781|H3-3B|H3.3B|H3F3B|H3-3A
- H3F3|H3F3A|PP781|H3-3B|H3.3B|H3F3B|H3-3A
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR23526-89
- EPR23526-89
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 15328 Da
- 15328 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Storage Temperature:
- Store at 4 °C
- Store at 4 °C
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- Dot, Flow Cyt (Intra), I-ELISA, ICC/IF, IHC-P
- Dot, Flow Cyt (Intra), I-ELISA, ICC/IF, IHC-P