Anti-RPL24 Rabbit Polyclonal Antibody
Catalog # ABCA_AB126172-50UL
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:Large ribosomal subunit protein eL24
- Antigen Symbol:RPL24
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Antigen Synonyms:60S ribosomal protein L24|60S ribosomal protein L30
- Molecular Weight:17779 Da
- Storage Temperature:Shipped at +4 °C. Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Recombinant Fragment Protein within Human 60S ribosomal protein L24 aa 1-150. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to RPL24.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RPL24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human