Anti-GSTA1 Rabbit Monoclonal Antibody [clone: EPR19969]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR19969] to GSTA1 - BSA and Azide free.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: GSTA1
Clonality: Monoclonal
Clone: EPR19969
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ABCA_AB251473-1MG
- ABCA_AB251473-100U
- Antigen Symbol:
- GSTA1
- GSTA1
- Antigen Name:
- Glutathione S-transferase A1
- Glutathione S-transferase A1
- Antigen Synonyms:
- 13-hydroperoxyoctadecadienoate peroxidase|GST HA subunit 1|GST class-alpha member 1|GST-epsilon|GSTA1-1|GTH1
- 13-hydroperoxyoctadecadienoate peroxidase|GST HA subunit 1|GST class-alpha member 1|GST-epsilon|GSTA1-1|GTH1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR19969
- EPR19969
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 25631 Da
- 25631 Da
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IP
- IP