Anti-USP47 Rabbit Polyclonal Antibody
ABCA_AB72143-100UL
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 47
- Antigen Symbol:USP47
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Deubiquitinating enzyme 47|Ubiquitin thioesterase 47|Ubiquitin-specific-processing protease 47
- Storage Buffer:BSA, Tris buffered saline
- Molecular Weight:157311 Da
- Storage Temperature:Shipped at +4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:0.2 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Synthetic Peptide within Human USP47. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IP
- Purification:Immunogen Affinity purification
- Packaging:Vial
Rabbit polyclonal to USP47.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: USP47
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human