Anti-ACADM/MCAD Rabbit Monoclonal Antibody [clone: EPR3708] (HRP (Horseradish Peroxidase))
ABCA_AB203465-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:MCAD
- Antigen Symbol:ACADM/MCAD
- Clonality:Monoclonal
- Clone:EPR3708
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Medium chain acyl-CoA dehydrogenase|MCADH|ACADM
- Molecular Weight:46588 Da
- Storage Temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Stable for 12 months at −20 °C. Store in the dark.
- Concentration:0.5 mg/ml
- Shipping Temperature:4 °C
- Immunogen:The exact immunogen used to generate this antibody is proprietary information
- Tested Applications:IHC-P
- Purification:Protein A affinity purification
- Packaging:Vial
HRP Rabbit monoclonal [EPR3708] to ACADM/MCAD.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ACADM/MCAD
Clonality: Monoclonal
Clone: EPR3708
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human