Anti-COX1/Cyclooxygenase 1 Rabbit Monoclonal Antibody [clone: EPR5866]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR5866] to COX1 / Cyclooxygenase 1.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: COX1 / Cyclooxygenase 1
Clonality: Monoclonal
Clone: EPR5866
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Specifications
- Catalog No:
- ABCA_AB109025-20UL
- ABCA_AB109025-1ML
- ABCA_AB109025-100U
- Antigen Symbol:
- COX1 / Cyclooxygenase 1
- COX1 / Cyclooxygenase 1
- COX1 / Cyclooxygenase 1
- Antigen Name:
- COX1
- COX1
- COX1
- Antigen Synonyms:
- PTGS1|Prostaglandin G/H synthase 1|Cyclooxygenase-1|Prostaglandin H2 synthase 1|Prostaglandin-endoperoxide synthase 1|COX-1|PGH synthase 1|PGHS-1|PHS 1
- PTGS1|Prostaglandin G/H synthase 1|Cyclooxygenase-1|Prostaglandin H2 synthase 1|Prostaglandin-endoperoxide synthase 1|COX-1|PGH synthase 1|PGHS-1|PHS 1
- PTGS1|Prostaglandin G/H synthase 1|Cyclooxygenase-1|Prostaglandin H2 synthase 1|Prostaglandin-endoperoxide synthase 1|COX-1|PGH synthase 1|PGHS-1|PHS 1
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- EPR5866
- EPR5866
- EPR5866
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Liquid
- Molecular Weight:
- 68686 Da
- 68686 Da
- 68686 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Protein A Affinity purification
- Storage Buffer:
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- BSA, Glycerol, Tissue culture supernatant, Sodium chloride, Tris glycine
- Storage Temperature:
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze/thaw cycle.
- Shipping Temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Shipped at +4 °C
- Tested Applications:
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P
- Flow Cyt (Intra), ICC/IF, IHC-P