Anti-DIRAS1 Rabbit Polyclonal Antibody
Catalog # ABCA_AB65139-100UG
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:GBTS1
- Antigen Symbol:DIRAS1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RIG|GTP-binding protein Di-Ras1|Distinct subgroup of the Ras family member 1|Ras-related inhibitor of cell growth|Small GTP-binding tumor suppressor 1
- Storage Buffer:PBS, Glycerol, Sodium chloride
- Molecular Weight:22329 Da
- Storage Temperature:Shipped at +4 °C. Store at –20 °C. Stable for 12 months at –20 °C.
- Concentration:1 mg/ml
- Shipping Temperature:+4 °C
- Immunogen:Synthetic Peptide within Human DIRAS1 aa 100 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to DIRAS1.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DIRAS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human