Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CISD1/MitoNEET Rabbit Monoclonal Antibody [clone: EPR29116-27]
Anti-CISD1/MitoNEET Rabbit Monoclonal Antibody [clone: EPR29116-27]
Supplier:  Abcam
New Product
Certificates
Anti-CISD1/MitoNEET Rabbit Monoclonal Antibody [clone: EPR29116-27]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCA_AB318199-100U
  • ABCA_AB318199-20UL
  • ABCA_AB318199-1ML
  • Antigen Symbol:
  • CISD1/MitoNEET
  • CISD1/MitoNEET
  • CISD1/MitoNEET
  • Antigen Name:
  • C10orf70
  • C10orf70
  • C10orf70
  • Antigen Synonyms:
  • ZCD1|MDS029|CISD1|CDGSH iron-sulfur domain-containing protein 1|Cysteine transaminase CISD1|MitoNEET
  • ZCD1|MDS029|CISD1|CDGSH iron-sulfur domain-containing protein 1|Cysteine transaminase CISD1|MitoNEET
  • ZCD1|MDS029|CISD1|CDGSH iron-sulfur domain-containing protein 1|Cysteine transaminase CISD1|MitoNEET
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR29116-27
  • EPR29116-27
  • EPR29116-27
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • The exact immunogen used to generate this antibody is proprietary information
  • Form:
  • Liquid
  • Liquid
  • Liquid
  • Molecular Weight:
  • 12199 Da
  • 12199 Da
  • 12199 Da
  • Purification:
  • Protein A affinity purification
  • Protein A affinity purification
  • Protein A affinity purification
  • Storage Temperature:
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle.
  • Shipping Temperature:
  • 4 °C
  • 4 °C
  • 4 °C
  • Tested Applications:
  • ICC/IF, IHC-P
  • ICC/IF, IHC-P
  • ICC/IF, IHC-P

Specifications