Anti-OB Cadherin Rabbit Monoclonal Antibody [clone: HL10044]
ABCA_AB289365-100U
: Abcam
New Product
- Antibody Type:Primary
- Antigen Name:Cadherin-11
- Antigen Symbol:OB Cadherin
- Clonality:Monoclonal
- Clone:HL10044
- Host:Rabbit
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:OSF-4|Osteoblast cadherin|OB-cadherin|CDH11
- Molecular Weight:87965 Da
- Storage Temperature:Store at 4 °C
- Shipping Temperature:4 °C
- Immunogen:Recombinant Fragment Protein within Human CDH11. The exact immunogen used to generate this antibody is proprietary information.
- Purification:Protein A affinity purification
- Packaging:Vial
Rabbit monoclonal [HL10044] to OB Cadherin - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: OB Cadherin
Clonality: Monoclonal
Clone: HL10044
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Cat