Anti-MAX Rabbit Polyclonal Antibody
Catalog # ABCA_AB101271-100U
Supplier: Abcam
New Product
Specifications
- Antibody Type:Primary
- Antigen Name:BHLHD4
- Antigen Symbol:MAX
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Protein max|Class D basic helix-loop-helix protein 4|Myc-associated factor X
- Storage Buffer:Tris citrate/phosphate
- Molecular Weight:18275 Da
- Storage Temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human MAX aa 100 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested Applications:IHC-P
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to MAX.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MAX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human