Anti-delta Sarcoglycan Rabbit Monoclonal Antibody [clone: EPR8706]
: Abcam
New Product
Rabbit monoclonal [EPR8706] to delta Sarcoglycan - BSA and Azide free.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: delta Sarcoglycan
Clonality: Monoclonal
Clone: EPR8706
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ABCA_AB240108-100U
- ABCA_AB240108-1MG
- Antigen Symbol:
- delta Sarcoglycan
- delta Sarcoglycan
- Antigen Name:
- Delta-sarcoglycan
- Delta-sarcoglycan
- Antigen Synonyms:
- Delta-SG|35 kDa dystrophin-associated glycoprotein|35DAG|SGCD
- Delta-SG|35 kDa dystrophin-associated glycoprotein|35DAG|SGCD
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR8706
- EPR8706
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information
- The exact immunogen used to generate this antibody is proprietary information
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 32071 Da
- 32071 Da
- Purification:
- Protein A affinity purification
- Protein A affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Temperature:
- Store at 4 °C. Do not freeze.
- Store at 4 °C. Do not freeze.
- Shipping Temperature:
- 4 °C
- 4 °C
- Tested Applications:
- IHC-P, mIHC
- IHC-P, mIHC