Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MGMT Rabbit Monoclonal Antibody [clone: EPR4397] (Alexa Fluor® 488)
  ABCA_AB318574-100U
 :  Abcam
New Product
undefined
Anti-MGMT Rabbit Monoclonal Antibody [clone: EPR4397] (Alexa Fluor® 488)
  ABCA_AB318574-100U
 :  Abcam
 :  AB318574-100UL
New Product

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Methylated-DNA--protein-cysteine methyltransferase
  • Antigen Symbol:
    MGMT
  • Clonality:
    Monoclonal
  • Clone:
    EPR4397
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    100 µl
  • Form:
    Liquid
  • Antigen Synonyms:
    6-O-methylguanine-DNA methyltransferase|O-6-methylguanine-DNA-alkyltransferase
  • Molecular Weight:
    21646 Da
  • Storage Temperature:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C. Avoid freeze/thaw cycle. Store in the dark.
  • Concentration:
    0.5 mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    The exact immunogen used to generate this antibody is proprietary information
  • Tested Applications:
    Antibody labelling, target binding affinity
  • Purification:
    Protein A affinity purification
  • Packaging:
    Vial

 

 

Alexa Fluor® 488 Rabbit monoclonal [EPR4397] to MGMT.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MGMT
Clonality: Monoclonal
Clone: EPR4397
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: